Learn More
Smooth Muscle Actic Specific Rabbit anti-Human, Unconjugated, Clone: 5H7, Proteintech
Rabbit Recombinant Antibody
140.25 CHF - 413.21 CHF
Spezifikation
| Antigen | Smooth Muscle Actic Specific |
|---|---|
| Klon | 5H7 |
| Konzentration | 260 μg/mL |
| Anwendungen | Western Blot, Immunohistochemistry, Immunofluorescence |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16820436
|
Proteintech
80008-1-RR-20UL |
20 μL |
140.25 CHF
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16810436
|
Proteintech
80008-1-RR-100UL |
100 μL |
413.21 CHF
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The smooth muscle actin specific antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human smooth muscle actin specific. This antibody recognizes human, mouse, rat, chicken, pig antigen. The smooth muscle actin specific antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
| Smooth Muscle Actic Specific | |
| 260 μg/mL | |
| Monoclonal | |
| Rabbit | |
| 59 | |
| Primary |
| 5H7 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Human, Mouse, Rat, Chicken, Pig | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.