Learn More
Smooth Muscle Actic Specific Rabbit anti-Human, Unconjugated, Clone: 5H7, Proteintech
Rabbit Recombinant Antibody
Marke:  Proteintech 80008-1-RR-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16820436
  • 140.25 CHF / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The smooth muscle actin specific antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human smooth muscle actin specific. This antibody recognizes human, mouse, rat, chicken, pig antigen. The smooth muscle actin specific antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
Spezifikation
Smooth Muscle Actic Specific
Monoclonal
260 μg/mL
Rabbit
Primary
Human, Mouse, Rat, Chicken, Pig
Western Blot, Immunohistochemistry, Immunofluorescence
5H7
Unconjugated
20 μL
59