Learn More
Smooth Muscle Actic Specific Rabbit anti-Human, Unconjugated, Clone: 5H7, Proteintech
Rabbit Recombinant Antibody
Marke: Proteintech 80008-1-RR-100UL
Beschreibung
The smooth muscle actin specific antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human smooth muscle actin specific. This antibody recognizes human, mouse, rat, chicken, pig antigen. The smooth muscle actin specific antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
Smooth Muscle Actic Specific | |
Monoclonal | |
260 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat, Chicken, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence | |
5H7 | |
Unconjugated | |
100 μL | |
59 | |