missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP130 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48 CHF - 385.92 CHF
Spezifikation
| Antigen | SAP130 |
|---|---|
| Konzentration | 0.13 mg/mL |
| Anwendungen | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|
16819663
|
Proteintech
12130-1-AP-20UL |
20 μL |
136.48 CHF
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16809663
|
Proteintech
12130-1-AP-150UL |
150 μL |
385.92 CHF
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Description
3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.Specifications
| SAP130 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q8BIH0, Q9H0E3 | |
| 269003, 307527, 79595 | |
| SAP130 Fusion Protein Ag2774 | |
| Primary | |
| -20°C | |
| SAP130 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| SAP130 | |
| SAP130 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title