missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK9/Cyclin T1 Recombinant Human Protein
CDK9 is coexpressed and copurified with CyclinT1.
2895.00 CHF - 18120.00 CHF
Spezifikation
Zugriffsnummer | NP_001252 I NP_001231 |
---|---|
Zur Verwendung mit (Anwendung) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Zusammensetzung | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, and50% Glycerol |
Molekulargewicht | 131.7kDa |
Lagerungsbedingungen | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15418613
|
Invitrogen™
PR7541B |
100μg |
2895.00 CHF
100 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
10620385
|
Invitrogen™
PV4280 |
1mg |
18120.00 CHF
1 Milligramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
CDK9 is a serine/threonine kinase which when complexed with CyclinT1 forms an active holoenzyme complex. This complex plays a key role in regulating the eukaryotic cell cycle.
Spezifikation
NP_001252 I NP_001231 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, and50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CDK and CHK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
131.7kDa | |
RUO - research use only | |
CDK9/Cyclin T1 | |
Human | |
His Tag | |
Frozen |