Learn More
Invitrogen™ CDK9/Cyclin T1 Recombinant Human Protein
CDK9 is coexpressed and copurified with CyclinT1.
Marke: Invitrogen™ PR7541B
Additional Details : Gewicht : 0.01000kg
Beschreibung
CDK9 is a serine/threonine kinase which when complexed with CyclinT1 forms an active holoenzyme complex. This complex plays a key role in regulating the eukaryotic cell cycle.
Spezifikation
NP_001252 I NP_001231 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, and50% Glycerol | |
100μg | |
RUO - research use only | |
CDK9/Cyclin T1 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
131.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CDK and CHK Kinase Family | |
Recombinant | |
Baculovirus |