missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDK9/Cyclin T1 Recombinant Human Protein

CDK9 is coexpressed and copurified with CyclinT1.

Marke:  Invitrogen™ PV4280

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 10620385

  • 18120.00 CHF / 1 Milligramm
Verfügbar ab: 23-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

CDK9 is a serine/threonine kinase which when complexed with CyclinT1 forms an active holoenzyme complex. This complex plays a key role in regulating the eukaryotic cell cycle.

Spezifikation

Spezifikation

NP_001252 I NP_001231
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, and50% Glycerol
1mg
RUO - research use only
CDK9/Cyclin T1
Human
His Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
131.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
CDK and CHK Kinase Family
Recombinant
Baculovirus
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote