missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Mouse anti-Human,Baboon,Cynomolgus,Pig,Rhesus Ma, BB700, Clone: B-ly4, BD Horizon™
Mouse Monoclonal Antibody
810.00 CHF
Spezifikation
Antigen | CD21 |
---|---|
Klon | B-ly4 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Brilliant Blue 700 (BB700) |
Beschreibung
CD21 Monoclonal antibody specifically detects CD21 in Human,Baboon,Cynomolgus,Pig,Rhesus Ma samples. It is validated for Flow Cytometry.Spezifikation
CD21 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Mouse | |
Human, Baboon, Cynomolgus, Pig, Rhesus Macaque | |
Primary |
B-ly4 | |
Monoclonal | |
Purified | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |