Learn More
CD21 Mouse anti-Human,Baboon,Cynomolgus,Pig,Rhesus Ma, BB700, Clone: B-ly4, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566569
Additional Details : Gewicht : 0.00840kg
Beschreibung
CD21 Monoclonal antibody specifically detects CD21 in Human,Baboon,Cynomolgus,Pig,Rhesus Ma samples. It is validated for Flow Cytometry.Spezifikation
CD21 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
B-ly4 | |
Mouse | |
RUO | |
Human, Baboon, Cynomolgus, Pig, Rhesus Macaque | |
Purified |