missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
ZNRF1 Polyclonal antibody specifically detects ZNRF1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZNRF1 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500-1:2000 |
| Zusammensetzung | PBS (pH 7.3), 50% glycerol |
| Gen-Alias | DKFZp434E229, EC 6.3.2, EC 6.3.2.-, FLJ14846, MGC15430, nerve injury gene 283, Nerve injury-induced gene 283 protein, nin283, NIN283E3 ubiquitin-protein ligase ZNRF1, zinc and ring finger 1, zinc and ring finger protein 1, Zinc/RING finger protein 1 |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human ZNRF1 (NP_115644.1). MGGKQSTAARSRGPFPGVSTDDSAVPPPGGAPHFGHYRTGGGAMGLRSRSVSSVAGMGMDPSTAGGVPFGLYTPASRGTGDSERAPGGGGSASDSTYAHGNGYQETGGGHHRDGMLYLGSRASLADALPLHIAPRWFSSHSGFKCPICSKSVASDEMEMHFIMCLSKPRL |
| Reinigungsverfahren | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?