missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF622 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20804-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ZNF622 may behave as an activator of the bound transcription factor, MYBL2, and be involved in embryonic development.Spezifikation
ZNF622 | |
Polyclonal | |
Unconjugated | |
ZNF622 | |
Zinc finger like protein 9, zinc finger protein 622, ZNF622, ZPR9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294846, 52521, 90441 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TM96, Q91VY9, Q969S3 | |
Znf622 | |
ZNF622 Fusion Protein Ag14741 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |