Learn More
ZEB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21544-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The calculated molecular weight of ZEB1 is 124 kDa, but the post phosphorylation of protein is about 190-210 kDa. This is a rabbit polyclonal antibody raised against part of the full length of ZEB1 of human.
THOC3 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.TEX1 is part of the TREX (transcription/export) complex, which includes THO2 (MIM 300395), HPR1 (MIM 606930), ALY (MIM 604171), and UAP56 (MIM 606390).Spezifikation
ZEB1 | |
Polyclonal | |
Unconjugated | |
ZEB1 | |
AREB6, BZP, DELTA EF1, delta EF1, Negative regulator of IL2, NIL 2 A, NIL 2 A zinc finger protein, NIL 2A, NIL2A, TCF 8, TCF8, Transcription factor 8, ZEB, ZEB1, ZFHEP, ZFHX1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21417, 25705, 6935 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P37275, Q62947, Q64318 | |
ZEB1 | |
ZEB1 Fusion Protein Ag16204 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |