Learn More
WSB1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11666-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
WSB1 (WD repeat and SOCS box-containing 1), also known as SWIP1, is a 421 amino acid protein that contains one SOCS box domain and six WD repeats. Expressed as multiple alternatively spliced isoforms, WSB1 is thought to function as part of an SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex that mediates the ubiquitination and proteasomal degradation of target proteins, such as DIO2. Overexpression of WSB1 is implicated in pancreatic cancer progression, suggesting a role for WSB1 in carcinogenesis.Spezifikation
WSB1 | |
Polyclonal | |
Unconjugated | |
WSB1 | |
SWIP1, WSB 1, WSB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26118 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y6I7 | |
WSB1 | |
WSB1 Fusion Protein Ag2211 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |