Learn More
Willin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21039-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Although willin is found in the cytoplasm and membrane, it localizes to the nuclei of almost all tumors. This antibody was raised against the C-terminal region of human FRMD6 protein and is expected to recognize the endogenous FRMD6.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Spezifikation
Willin | |
Polyclonal | |
Unconjugated | |
FRMD6 | |
EX1, FERM domain containing 6, FRMD6, Willin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122786, 257646, 319710 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C0V9, Q8VII0, Q96NE9 | |
Frmd6 | |
Willin Fusion Protein Ag15375 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |