missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11558-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
WFS1 is a novel component of Wolfram syndrome, a rare form of juvenile diabetes. WFS1 plays an important role in maintaining homeostasis of the endoplasmic reticulum (ER) in the pancreas. It is normally up-regulated during insulin secretion, whereas inactivation of the protein can cause ER stress. Chronic ER stress is a major involvement in Wolfram syndrome.Spezifikation
WFS1 | |
Polyclonal | |
Unconjugated | |
WFS1 | |
WFRS, WFS, WFS1, Wolfram syndrome 1 (wolframin), WOLFRAMIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22393, 7466 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O76024, P56695 | |
WFS1 | |
WFS1 Fusion Protein Ag2114 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |