Learn More
WDR91 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 27424-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The WDR91 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WDR91. This antibody recognizes human antigen. The WDR91 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| WDR91 | |
| 750 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| HSPC049, WD repeat domain 91, WDR91 | |
| 150 μL | |
| 29062 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |