missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VN1R4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18846-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Putative pheromone receptor. Various VN1R4 alleles are known. T The chimpanzee and gorilla orthologous proteins do not exist, their genes are pseudogenes.Spezifikation
VN1R4 | |
Polyclonal | |
Unconjugated | |
VN1R4 | |
hGPCR27, V1r like receptor 4, V1RL4, VN1R4, vomeronasal 1 receptor 4, Vomeronasal type 1 receptor 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
317703 | |
-20°C | |
Liquid |
Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7Z5H5 | |
VN1R4 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |