missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
265.00 CHF - 532.00 CHF
Spezifikation
| Antigen | HIV-1 Gag p24 |
|---|---|
| Klon | 2230C |
| Verdünnung | Western Blot 0.1 ug/mL |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18775039
![]() |
R&D Systems
MAB9539-SP |
25 μg |
265.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18791808
![]() |
R&D Systems
MAB9539-100 |
100 μg |
532.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
HIV-1 Gag p24 Monoclonal specifically detects HIV-1 Gag p24 in Viral samples. It is validated for Western Blot.Spezifikation
| HIV-1 Gag p24 | |
| Western Blot 0.1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 155030 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2230C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Q77YG1 | |
| E. coli-derived recombinant viral HIV-1 Gag p24 Pro149-Gly354 Accession # Q77YG1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects viral HIV-1 Gag p24 in Western blots. |
RUO
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts