Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Marke:  R&D Systems MAB9539-SP
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18775039
  • 265.00 CHF / 25 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
HIV-1 Gag p24 Monoclonal specifically detects HIV-1 Gag p24 in Viral samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
HIV-1 Gag p24
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Rabbit
Protein A or G purified from cell culture supernatant
RUO
155030
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG
Western Blot
2230C
Western Blot 0.1 ug/mL
Q77YG1
E. coli-derived recombinant viral HIV-1 Gag p24 Pro149-Gly354 Accession # Q77YG1
25 μg
Primary
Detects viral HIV-1 Gag p24 in Western blots.
Virus
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen