Learn More
Donkey anti-Rat IgG (H+L) Secondary Antibody, DyLight 650 (Pre-adsorbed), Novus Biologicals™
Artikelnummer. 18026995 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18026995 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18026995 Lieferant Novus Biologicals Lieferanten-Nr. NBP175663
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Donkey Polyclonal Antibody
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Flow Cytometry, Immunomicroscopy.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen IgG (H+L)
Anwendungen Flow Cytometry, Immunomicroscopy
Klassifikation Polyclonal
Konzentration LYOPH
Konjugat DyLight 650
Verdünnung Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Wirtsspezies Donkey
Immunogen Purified Rat IgG (H&L)
Reinigungsverfahren Affinity purified
Menge 1 mg
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Immunology
Primär oder sekundär Secondary
Testspezifität Based on IEP, this antibody reacts with heavy gamma chains on rat IgG and light chains on all rat immunoglobulins. This antibody has been pre-adsorbed against bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rabbit and sheep IgG.
Rekonstitution Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Zielspezies Rat
Inhalt und Lagerung Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Isotype IgG
Mehr anzeigen Weniger anzeigen
For Research Use Only