Learn More
VEGFR2/KDR/Flk-1 Antibody (89106R), PE/Atto594, Novus Biologicals™
Artikelnummer. 30036665 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30036665 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 30036665 Lieferant Novus Biologicals Lieferanten-Nr. FAB3572RPEATT594
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Monoclonal Antibody
VEGFR2/KDR/Flk-1 Monoclonal antibody specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen VEGFR2/KDR/Flk-1
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon 89106R
Konjugat PE/ATTO 594
Verdünnung Flow Cytometry
Zusammensetzung PBS
Gen-Alias CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2
Wirtsspezies Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGFR2/KDR/Flk-1, Ala20-Glu764, Accession # P35968
Reinigungsverfahren Protein A or G purified from cell culture supernatant
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
Primär oder sekundär Primary
Gen-ID (Entrez) 3791
Zielspezies Human
Inhalt und Lagerung Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1