missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11822-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
VAP5, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of vesicles and cell membranes. The VAMP5 gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. This VAMP family member may participate in vesicle trafficking events that are associated with myogenesis.Spezifikation
VAMP5 | |
Polyclonal | |
Unconjugated | |
VAMP5 | |
Myobrevin, VAMP 5, VAMP5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10791, 53620, 89818 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95183, Q9Z2J5, Q9Z2P8 | |
Vamp5 | |
VAMP5 Fusion Protein Ag2390 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |