Learn More
Use1/UBE2Z Antibody, Novus Biologicals™
Artikelnummer. 18677622 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
50 μg
Packungsgröße:
100 Mikroliter
50 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18677622 100 μg 100 Mikroliter
18644175 50 μg 50 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18677622 Lieferant Novus Biologicals Lieferanten-Nr. NBP297553100ul
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Polyclonal Antibody
Use1/UBE2Z Polyclonal antibody specifically detects Use1/UBE2Z in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Use1/UBE2Z
Anwendungen Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200
Zusammensetzung PBS (pH 7.0)
Gen-Alias MDS032, P31, protein p31, putative MAPK activating protein PM26, Putative MAPK-activating protein PM26, Q-SNARE, SLT1, SNARE-like tail-anchored protein 1 homolog, unconventional SNARE in the ER 1 homolog (S. cerevisiae), USE1L, USE1-like protein, vesicle transport protein USE1
Wirtsspezies Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Use1/UBE2Z fragment.
Reinigungsverfahren Antigen and protein A Affinity-purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Apoptosis, Cell Biology, Cell Cycle and Replication
Primär oder sekundär Primary
Gen-ID (Entrez) 55850
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG