missing translation for 'onlineSavingsMsg'
Learn More

URG4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 11998-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16839443

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 21-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The specific function of this protein remains unknown. URG4 is upregulated in the presence of hepatitis B virus (HBV)-encoded X antigen (HBxAg) and may contribute to the development of hepatocellular carcinoma by promoting hepatocellular growth and survival (Tufan et al., 2002 ).
Spezifikation

Spezifikation

URG4
Polyclonal
Unconjugated
URGCP
KIAA1507, up regulated gene 4, URG4, URGCP
Rabbit
Antigen Affinity Chromatography
RUO
55665
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8TCY9
URGCP
URG4 Fusion Protein Ag2630
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote