missing translation for 'onlineSavingsMsg'
Learn More
Learn More
URG4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11998-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The specific function of this protein remains unknown. URG4 is upregulated in the presence of hepatitis B virus (HBV)-encoded X antigen (HBxAg) and may contribute to the development of hepatocellular carcinoma by promoting hepatocellular growth and survival (Tufan et al., 2002 ).Spezifikation
URG4 | |
Polyclonal | |
Unconjugated | |
URGCP | |
KIAA1507, up regulated gene 4, URG4, URGCP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55665 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TCY9 | |
URGCP | |
URG4 Fusion Protein Ag2630 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |