missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC5D Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20241-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody is specific to UNC5D.
UNC5D, also named as KIAA1777 and UNC5H4, belongs to the unc-5 family. It is a receptor for netrin. It may be involved in axon guidance by mediating axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand.Spezifikation
UNC5D | |
Polyclonal | |
Unconjugated | |
UNC5D | |
KIAA1777, Netrin receptor UNC5D, PRO34692, Protein unc 5 homolog 4, Protein unc 5 homolog D, unc 5 homolog D (C. elegans), UNC5D, Unc5h4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
137970, 210801 | |
Antibody | |
IgG |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UXZ4, Q8K1S2 | |
Unc5d | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Liquid |