missing translation for 'onlineSavingsMsg'
Learn More

ULBP-2 Antibody - Azide and BSA Free, R&D Systems™

Artikelnummer. 18609970 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0.02 mL
0.1 mL
Packungsgröße:
0.02 Milliliter
0.10 Milliliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18609970 0.02 mL 0.02 Milliliter
18687291 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18609970 Lieferant Novus Biologicals Lieferanten-Nr. NBP2945890.02ml
 Eingeschränkter Artikel
Verfügbar ab: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ULBP-2 Polyclonal specifically detects ULBP-2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULBP-2
Anwendungen Immunocytochemistry/Immunofluorescence, Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Zusammensetzung PBS (pH 7.3), 50% glycerol
Gen-Alias ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 26-217 of human ULBP-2 (NP_079493.1). GRADPHSLCYDITVIPKFRPGPRWCAVQGQVDEKTFLHYDCGNKTVTPVSPLGKKLNVTTAWKAQNPVLREVVDILTEQLRDIQLENYTPKEPLTLQARMSCEQKAEGHSSGSWQFSFDGQIFLLFDSEKRMWTTVHPGARKMKEKWENDKVVAMSFHYFSMGDCIGWLEDFLMGMDSTLEPSAGAPLAMSS
Reinigungsverfahren Affinity purified
Menge 0.02 mL
Regulatorischer Status RUO
Forschungsgebiet Immunology, Innate Immunity
Primär oder sekundär Primary
Gen-ID (Entrez) 80328
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at -20°C. Avoid freeze-thaw cycles.
Produkttyp Antibody
Form Purified
Isotype IgG
Produktlinie Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.