missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13109-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3′ coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
UBXN11 | |
Polyclonal | |
Unconjugated | |
UBXN11 | |
COA 1, PP2243, SOC, SOCI, Socius, UBX domain protein 11, UBXD5, UBXN11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192207, 67586, 91544 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5T124, Q8R512, Q9D572 | |
UBXN11 | |
UBXN11 Fusion Protein Ag3772 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |