missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | UBXN11 |
---|---|
Konzentration | 0.47 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16841164
|
Proteintech
13109-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 06-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16831164
|
Proteintech
13109-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 06-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3′ coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
UBXN11 | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q5T124, Q8R512, Q9D572 | |
192207, 67586, 91544 | |
UBXN11 Fusion Protein Ag3772 | |
Primary | |
-20°C | |
UBXN11 |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
COA 1, PP2243, SOC, SOCI, Socius, UBX domain protein 11, UBXD5, UBXN11 | |
UBXN11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |