missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTRAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12203-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of a superfamily of divalent cation-dependent phosphodiesterases. The encoded protein associates with CD40, tumor necrosis factor (TNF) receptor-75 and TNF receptor associated factors (TRAFs), and inhibits nuclear factor-kappa-B activation. This protein has sequence and structural similarities with APE1 endonuclease, which is involved in both DNA repair and the activation of transcription factors.Spezifikation
TTRAP | |
Polyclonal | |
Unconjugated | |
TDP2 | |
5 Tyr DNA phosphodiesterase, AD022, EAP2, EAPII, ETS1 associated protein 2, ETS1 associated protein II, TDP2, TTRAP, Tyr DNA phosphodiesterase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
498749, 51567, 56196 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95551, Q3T1H5, Q9JJX7 | |
TDP2 | |
TTRAP Fusion Protein Ag2846 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |