missing translation for 'onlineSavingsMsg'
Learn More

Triacetoxy(methyl)silan, 96 %, Thermo Scientific Chemicals

Artikelnummer. 11456856
Click to view available options
Menge:
10 g
50 g
250 g
Unit Size:
10 Gramm
250 Gramm
50 Gramm
This item is not returnable. View return policy

Product Code. 11456856

Brand: Thermo Scientific Alfa Aesar B23974.30

Bulk oder benutzerdefiniertes Format anfordern

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

CAS: 4253-34-3 | C7H12O6Si | 220.25 g/mol

Dieses Markenprodukt von Thermo Scientific Chemicals war ursprünglich Teil des Produktportfolios von Alfa Aesar. Einige Dokumente und Etiketteninformationen enthalten möglicherweise Angaben zur bisherigen Marke. Die ursprüngliche Produkt- oder Artikelnummer bei Alfa Aesar oder die SKU-Referenz wurde im Rahmen des Übergangs der Marke zu Thermo Scientific Chemicals nicht geändert.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4253-34-3
Summenformel C7H12O6Si
Molekulargewicht (g/mol) 220.25
MDL-Nummer MFCD00008694
InChI-Schlüssel TVJPBVNWVPUZBM-UHFFFAOYSA-N
Synonym methyltriacetoxysilane, methylsilanetriyl triacetate, triacetoxymethylsilane, triacetoxy methyl silane, silanetriol, methyl-, triacetate, silane, methyltriacetoxy, methylsilanetriol triacetate, methyltrihydroxysilane triacetate, apk 1 silane derivative, triacetoxy-methylsilane
PubChem CID 77929
SMILES CC(=O)O[Si](C)(OC(C)=O)OC(C)=O

Specifications

Siedepunkt 87°C to 88°C (3mmHg)
Chemischer Name oder Material Triacetoxy(methyl)silane
Schmelzpunkt 40°C to 45°C
Menge 250 g
UN-Nummer UN1759
Beilstein 1788668
Flammpunkt 85°C (185°F)
IUPAC-Name Bis(Acetyloxy)(Methyl)silylacetat
Formelmasse 220.25
Reinheit (%) 96%
Empfindlichkeit Moisture sensitive
Dichte 1.175
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.