Learn More
Abnova™ TREM2 Recombinant Protein
Human TREM2 full-length ORF recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00054209-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL)
- Theoretical MW (kDa): 50.16
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MEPLRLLILLFVTELSGAHNTTVFQGVAGQSLQVSCPYDSMKHWGRRKAWCRQLGEKGPCQRVVSTHNLWLLSFLRRWNGSTAITDDTLGGTLTITLRNLQPHDAGLYQCQSLHGSEADTLRKVLVEVLADPLDHRDAGDLWFPGESESFEDAHVEHSISRPSQGSHLPSCLSKEPLGRRNPLPTHFHPSPPGLHLSHQDSSSQRPLGCSLAWTEARDTSTQ
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH32362 | |
Solution | |
54209 | |
TREM2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TREM2 | |
Human | |
Yes |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
50.16 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MEPLRLLILLFVTELSGAHNTTVFQGVAGQSLQVSCPYDSMKHWGRRKAWCRQLGEKGPCQRVVSTHNLWLLSFLRRWNGSTAITDDTLGGTLTITLRNLQPHDAGLYQCQSLHGSEADTLRKVLVEVLADPLDHRDAGDLWFPGESESFEDAHVEHSISRPSQGSHLPSCLSKEPLGRRNPLPTHFHPSPPGLHLSHQDSSSQRPLGCSLAWTEARDTSTQ | |
TREM-2/Trem2a/Trem2b/Trem2c | |
TREM2 | |
Wheat Germ (in vitro) | |
GST |