missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trappc9/NIBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20287-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.Spezifikation
Trappc9/NIBP | |
Polyclonal | |
Unconjugated | |
TRAPPC9 | |
1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, KIAA1882, mKIAA1882, NIBP, Trappc9, TRS130 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
76510, 83696 | |
Antibody | |
IgG |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3U0M1, Q96Q05 | |
Trappc9 | |
Trappc9,NIBP Fusion Protein Ag14017 | |
20 μL | |
Primary | |
Mouse, Human | |
Liquid |