missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIL Mouse anti-Human, Clone: 1B9B4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66756-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Tumor necrosis factor receptor superfamily member 1A (TNFRSF1A) is a protein that in humans is encoded by the TNFRSF1A gene. The protein encoded by this gene is a member of the Tumor necrosis factor receptor superfamily, which also contains TNFRSF1B. The TNFR1 gene is mapped to 12pter-cen. it encodes a protein of 455 amino acids. This receptor can activate the transcription factor NF-KB, mediate apoptosis, and function as a regulator of inflammation.Spezifikation
TRAIL | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P50591 | |
TNFSF10 | |
TRAIL Fusion Protein Ag25746 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1B9B4 | |
Unconjugated | |
TNFSF10 | |
Apo 2 ligand, Apo 2L, APO2L, CD253, Protein TRAIL, TL2, TNFSF10, TRAIL | |
Mouse | |
Protein G | |
RUO | |
8743 | |
-20°C | |
Liquid |