missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPTE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12982-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.Spezifikation
TPTE | |
Polyclonal | |
Unconjugated | |
TPTE | |
Cancer/testis antigen 44, CT44, PTEN2, TPTE, Tumor antigen BJ HCC 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7179 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P56180 | |
TPTE | |
TPTE Fusion Protein Ag3606 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |