missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12479-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the sedolisin family of serine proteases. The protease functions in the lysosome to cleave N-terminal tripeptides from substrates, and has weaker endopeptidase activity. It is synthesized as a catalytically-inactive enzyme which is activated and auto-proteolyzed upon acidification. Mutations in this gene result in late-infantile neuronal ceroid lipofuscinosis, which is associated with the failure to degrade specific neuropeptides and a subunit of ATP synthase in the lysosome.Spezifikation
TPP1 | |
Polyclonal | |
Unconjugated | |
TPP1 | |
CLN2, GIG1, LPIC, TPP 1, TPP I, TPP1, Tripeptidyl aminopeptidase, Tripeptidyl peptidase 1, tripeptidyl peptidase I, UNQ267/PRO304 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1200, 12751, 83534 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14773, O89023, Q9EQV6 | |
Tpp1 | |
TPP1 Fusion Protein Ag3173 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |