missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMSB4X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19850-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes an actin sequestering protein which plays a role in regulation of actin polymerization. The protein is also involved in cell proliferation, migration, and differentiation. This gene escapes X inactivation and has a homolog on chromosome Y.Spezifikation
TMSB4X | |
Polyclonal | |
Unconjugated | |
TMSB4X | |
FX, PTMB4, Seraspenide, T beta 4, TB4X, THYB4, Thymosin beta 4, thymosin beta 4, X linked, TMSB4, TMSB4X | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19241, 7114, 81814 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20065, P62328, P62329 | |
TMSB4X | |
TMSB4X Fusion Protein Ag13914 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |