missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMSB4X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | TMSB4X |
---|---|
Konzentration | 0.23 mg/mL |
Anwendungen | Immunoprecipitation, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839334
|
Proteintech
19850-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829334
|
Proteintech
19850-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes an actin sequestering protein which plays a role in regulation of actin polymerization. The protein is also involved in cell proliferation, migration, and differentiation. This gene escapes X inactivation and has a homolog on chromosome Y.Spezifikation
TMSB4X | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P20065, P62328, P62329 | |
19241, 7114, 81814 | |
TMSB4X Fusion Protein Ag13914 | |
Primary | |
-20°C | |
TMSB4X |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FX, PTMB4, Seraspenide, T beta 4, TB4X, THYB4, Thymosin beta 4, thymosin beta 4, X linked, TMSB4, TMSB4X | |
TMSB4X | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |