missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMOD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11753-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody was raised against the full-length human TMOD4 and may cross-react with other Tmods.
TMOD4 blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.Spezifikation
TMOD4 | |
Polyclonal | |
Unconjugated | |
TMOD4 | |
SK TMOD, Skeletal muscle tropomodulin, TMOD4, Tropomodulin 4, tropomodulin 4 (muscle) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295261, 29765, 50874 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JLH8, Q9NZQ9 | |
Tmod4 | |
TMOD4 Fusion Protein Ag2349 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |