missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMOD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | TMOD4 |
---|---|
Konzentration | 0.17 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839043
|
Proteintech
11753-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829043
|
Proteintech
11753-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This antibody was raised against the full-length human TMOD4 and may cross-react with other Tmods.
TMOD4 blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.Spezifikation
TMOD4 | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q9JLH8, Q9NZQ9 | |
295261, 29765, 50874 | |
TMOD4 Fusion Protein Ag2349 | |
Primary | |
-20°C | |
TMOD4 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SK TMOD, Skeletal muscle tropomodulin, TMOD4, Tropomodulin 4, tropomodulin 4 (muscle) | |
Tmod4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |