Learn More
TMEM16K Antibody, Novus Biologicals™
Artikelnummer. 18298621 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18298621 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18298621 Lieferant Novus Biologicals Lieferanten-Nr. NBP159667
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Polyclonal Antibody
TMEM16K Polyclonal specifically detects TMEM16K in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen TMEM16K
Anwendungen Western Blot
Klassifikation Polyclonal
Konzentration 0.5 mg/ml
Konjugat Unconjugated
Verdünnung Western Blot 1.0 ug/ml
Zusammensetzung PBS, 2% Sucrose with 0.09% Sodium Azide
Gen-Zugriffsnummer Q9NW15
Gen-Alias anoctamin 10, FLJ10375, MGC47890, Transmembrane protein 16KTMEM16Kanoctamin-10
Gensymbole ANO10
Wirtsspezies Rabbit
Immunogen Synthetic peptides corresponding to TMEM16K(transmembrane protein 16K) The peptide sequence was selected from the C terminal of TMEM16K. Peptide sequence LKADIDATLYEQVILEKEMGTYLGTFDDYLELFLQFGYVSLFSCVYPLAA.
Molekulargewicht des Antigens 76 kDa
Reinigungsverfahren Affinity purified
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 55129
Rekonstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Zielspezies Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG