Learn More
TLR8 Antibody (44C143), Alexa Fluor™ 750, Novus Biologicals™
Artikelnummer. 18103952 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18103952 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18103952 Lieferant Novus Biologicals Lieferanten-Nr. NBP224917AF750
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Monoclonal Antibody
TLR8 Monoclonal specifically detects TLR8 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen TLR8
Anwendungen Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon 44C143
Konjugat Alexa Fluor 750
Verdünnung Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
Zusammensetzung 50 mM sodium borate with 0.05% sodium azide
Gen-Alias CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8
Gensymbole TLR8
Wirtsspezies Mouse
Immunogen This antibody was developed against a KLH-conjugated synthetic peptide of human TLR8, within amino acids 750-850.
Reinigungsverfahren Protein G purified
Menge 0.1 mL
Forschungsgebiet Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors
Primär oder sekundär Primary
Gen-ID (Entrez) 51311.0
Zielspezies Human, Mouse
Inhalt und Lagerung Store at 4°C in the dark.
Isotype IgG1 κ