missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TLR3 Antibody (CNTO5429) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
248.00 CHF - 1080.00 CHF
Spezifikation
| Antigen | TLR3 |
|---|---|
| Klon | CNTO5429 |
| Anwendungen | Flow Cytometry, ELISA, Functional Assay |
| Klassifikation | Monoclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30045998
|
Novus Biologicals™
NBP3-28194-50ug |
50 μg |
248.00 CHF
50 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30047287
|
Novus Biologicals™
NBP3-28194-100ug |
100 μg |
379.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30048547
|
Novus Biologicals™
NBP3-28194-1mg |
1 mg |
1080.00 CHF
1 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
TLR3 Monoclonal antibody specifically detects TLR3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpezifikation
| TLR3 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| TLR3/ CD283 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| CNTO5429 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| O15455 | |
| 7098 | |
| IgG2 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts