missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11372-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.Spezifikation
TLE3 | |
Polyclonal | |
Unconjugated | |
TLE3 | |
ESG, ESG3, GRG3, HsT18976, KIAA1547, TLE3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21887, 7090, 84424 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q04726, Q08122, Q9JIT3 | |
TLE3 | |
TLE3 Fusion Protein Ag1930 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |