Learn More
TIP30 Antibody (5D6), Alexa Fluor™ 488, Novus Biologicals™
Artikelnummer. 18285750 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18285750 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18285750 Lieferant Novus Biologicals Lieferanten-Nr. NBP259482AF488
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Monoclonal Antibody
TIP30 Monoclonal antibody specifically detects TIP30 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen TIP30
Anwendungen Western Blot, ELISA, Immunofluorescence
Klassifikation Monoclonal
Klon 5D6
Konjugat Alexa Fluor 488
Zusammensetzung 50mM Sodium Borate
Gen-Alias 30 kDa HIV-1 TAT-interacting protein, CC3oxidoreductase HTATIP2, EC 1.1.1, EC 1.1.1.-, FLJ26963, HIV-1 Tat interactive protein 2, 30 kDa, HIV-1 Tat interactive protein 2, 30kDa, HIV-1 TAT-interactive protein 2, SDR44U1, Tat-interacting protein (30kD), TIP30short chain dehydrogenase/reductase family 44U, member 1
Wirtsspezies Mouse
Immunogen Recombinant human TIP30 (1-242aa) purified from E. coli
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Apoptosis
Primär oder sekundär Primary
Gen-ID (Entrez) 10553
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ