missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TINP1 Mouse anti-Human, Clone: 1E3B11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66374-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. Mice with a mutation that results in reduced expression of the ortholog of this protein are retarded for growth, develop severe tremors by 2 to 3 weeks of age followed by hindlimb paralysis and death by 6 to 10 weeks of age. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Spezifikation
TINP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95478 | |
NSA2 | |
TINP1 Fusion Protein Ag8822 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1E3B11 | |
Unconjugated | |
NSA2 | |
CDK105, Hairy cell leukemia protein 1, HCL G1, HUSSY29, NSA2, TINP1, YR 29 | |
Mouse | |
Protein G | |
RUO | |
10412 | |
-20°C | |
Liquid |