missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TEC, His Tag Recombinant Protein
Marke: Invitrogen PV4090
Additional Details : Gewicht : 0.00900kg
Beschreibung
- A non-receptor protein-tyrosine kinase containing a pleckstrin homology domain
- It is highly expressed in many hematopoietic cell lines
Spezifikation
NP_003206 | |
76.4 kDa | |
TEC, PSCTK4 | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Frozen |
Kinase Assay | |
1 mg | |
TEC Kinase Family | |
Recombinant | |
Storage Buffer: 20 mM Tris, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.05% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Full Length | |
Drug Development |