missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12447-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Knockout studies in mice indicate that this gene is important for specification of paraxial mesoderm structures.Spezifikation
TBX6 | |
Polyclonal | |
Unconjugated | |
TBX6 | |
DFNB67, T box 6, T box protein 6, TBX6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21389, 365371, 6911 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D3ZJK7, O95947, P70327 | |
Tbx6 | |
TBX6 Fusion Protein Ag3050 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |