missing translation for 'onlineSavingsMsg'
Learn More

TBX6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 12447-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16850164

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 24-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Knockout studies in mice indicate that this gene is important for specification of paraxial mesoderm structures.
Spezifikation

Spezifikation

TBX6
Polyclonal
Unconjugated
TBX6
DFNB67, T box 6, T box protein 6, TBX6
Rabbit
Antigen Affinity Chromatography
RUO
21389, 365371, 6911
-20°C
Liquid
Western Blot, Immunoprecipitation
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
D3ZJK7, O95947, P70327
Tbx6
TBX6 Fusion Protein Ag3050
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote