Learn More
Invitrogen™ Human TBK1, GST Tag Recombinant Protein
TBK1 serine/threonine kinase is a member of the IKK family and is involved in NF-kappa B activation and IRF3 signaling.
Marke: Invitrogen™ A31520
Additional Details : Gewicht : 0.00900kg
Beschreibung
This purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ assay technologies to identify novel kinase inhibitors. TBL1 is also available in our SelectScreen™ Kinase Profiling Service.
- Validated for use with the LanthaScreen & Z'-LYTE assay technology
- Available in multiple pack sizes for convenient use
- Also available in our SelectSceen Kinase Profiling Service
Spezifikation
AAH34950.1 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutath¡one, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. | |
1mg | |
RUO - research use only | |
NAK/T2K/FLJ11330 | |
Full Length | |
Recombinant | |
Insect |
Kinase Assay | |
109.9kDa | |
Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws. | |
Full Length, Recombinant, Active | |
CK1 Kinase Family | |
Human | |
GST Tag |